Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256071.2 | NP_001243000.2 | 8 | 1584 | ||
NM_020954.3 | NP_066005.2 | 8 | 1584 | |||
XM_005257545.4 | XP_005257602.2 | 9 | 1772 | |||
XM_005257546.4 | XP_005257603.2 | 9 | 1676 | |||
XM_006721995.3 | XP_006722058.1 | 9 | 2059 | |||
XM_011525084.2 | XP_011523386.1 | 9 | 1772 | |||
XM_011525086.2 | XP_011523388.1 | 9 | 1772 | |||
XM_011525087.2 | XP_011523389.1 | 9 | 1772 | |||
XM_017024905.1 | XP_016880394.1 | 4 | 639 | |||
GenBank mRNA | AB046838.1 | - | 5 | 1029 | ||
AB537889.1 | - | 8 | 1577 | |||
AL832920.1 | - | 8 | 1571 | |||
BC036891.1 | - | 8 | 1600 | 5501834 | ||
BC040341.1 | - | 8 | 1575 | 3873001 |