Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTHFSD SIRNA Assays ›
Gene Symbol:
MTHFSD
Gene Name:
methenyltetrahydrofolate synthetase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.16: 86530176 - 86555235 on Build GRCh38
UniGene ID:
Hs.343627
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS127748
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTHFSD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159377.1
NP_001152849.1
2
87
NM_001159378.1
NP_001152850.1
2
87
NM_001159379.1
NP_001152851.1
2
84
NM_001159380.1
NP_001152852.1
2
51
NM_022764.2
NP_073601.2
2
84
NR_027489.1
2
80
NR_027490.1
2
87
XM_005256101.1
XP_005256158.1
2
127
XM_005256105.1
XP_005256162.1
2
133
XM_005256106.1
XP_005256163.1
2
42
XM_011523280.2
XP_011521582.1
2
127
XM_011523282.2
XP_011521584.1
2
48
XM_011523283.2
XP_011521585.1
2
48
XM_011523284.2
XP_011521586.1
2
87
XM_011523285.1
XP_011521587.1
2
48
XM_011523286.1
XP_011521588.1
2
148
XM_011523287.1
XP_011521589.1
2
52
XM_011523289.1
XP_011521591.1
2
148
XM_017023571.1
XP_016879060.1
2
87
XM_017023572.1
XP_016879061.1
2
84
XM_017023573.1
XP_016879062.1
2
41
XM_017023574.1
XP_016879063.1
2
87
GenBank mRNA
AK023060.1
-
2
62
AK023955.1
-
2
41
AK096836.1
-
2
80
AK297726.1
-
2
84
BC013911.2
-
1
18
BC112057.1
-
2
67
8327525
BC112059.1
-
2
67
8327527
BC143702.1
-
2
52
9052216
BC143704.1
-
2
52
BC143705.1
-
2
51
BC143707.1
-
2
55
9052221
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
git-url: https://github.com/thermofisher/magellan-search
git-branch: integration/2.33.0-meta-desc