Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024938.3 | NP_001020109.1 | 4, 5 | 672 | ||
NM_001024939.3 | NP_001020110.1 | 4, 5 | 560 | |||
NM_001282864.1 | NP_001269793.1 | 4, 5 | 915 | |||
NM_030807.4 | NP_110434.3 | 5, 6 | 783 | |||
NR_104247.1 | 4, 5 | 668 | ||||
NR_104248.1 | 5, 6 | 783 | ||||
GenBank mRNA | AB209301.1 | - | 3, 4 | 318 | ||
AF443201.1 | - | 5, 6 | 614 | |||
AF479409.1 | - | 3, 4 | 353 | |||
AJ271290.1 | - | 4, 5 | 503 | |||
AK075467.1 | - | 4, 5 | 448 | |||
AK296169.1 | - | 4, 5 | 437 | |||
AK300196.1 | - | 4, 5 | 915 | |||
AK304409.1 | - | 4, 5 | 538 | |||
AK308589.1 | - | 4, 5 | 534 | |||
AK308602.1 | - | 4, 5 | 437 | |||
AK314502.1 | - | 5, 6 | 554 | |||
AK315937.1 | - | 4, 5 | 439 | |||
AK316049.1 | - | 4, 5 | 505 | |||
BC063525.1 | - | 5, 6 | 540 | |||
BC094735.1 | - | 4, 5 | 489 | 5304025 | ||
BC100807.2 | - | 4, 5 | 418 | |||
BC100808.2 | - | 4, 5 | 418 | 40000757 | ||
BC100809.1 | - | 4, 5 | 418 | 40000760 | ||
BC100810.1 | - | 4, 5 | 414 | |||
CR456373.1 | - | 4, 5 | 449 |