Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282495.1 | NP_001269424.1 | 7 | 1168 | ||
NM_001282496.1 | NP_001269425.1 | 6 | 1200 | |||
NM_001282497.1 | NP_001269426.1 | 7 | 1365 | |||
NM_001282498.1 | NP_001269427.1 | 4 | 916 | |||
NM_001321800.1 | NP_001308729.1 | 6 | 1106 | |||
NM_001321801.1 | NP_001308730.1 | 7 | 1229 | |||
NM_001321802.1 | NP_001308731.1 | 7 | 1362 | |||
NM_001321803.1 | NP_001308732.1 | 7 | 1226 | |||
NM_001321805.1 | NP_001308734.1 | 6 | 1239 | |||
NM_024325.5 | NP_077301.4 | 6 | 1242 | |||
XM_017028062.1 | XP_016883551.1 | 6 | 1083 | |||
GenBank mRNA | AK055734.1 | - | 2 | 671 | ||
AK096911.1 | - | 6 | 1243 | |||
BC011862.2 | - | 1 | 412 | |||
BC065009.1 | - | 7 | 1165 | 6166526 |