Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024593.3 | NP_078869.1 | 2 | 283 | ||
XM_005251303.1 | XP_005251360.1 | 2 | 240 | |||
XM_011517589.1 | XP_011515891.1 | 2 | 240 | |||
XM_011517590.2 | XP_011515892.1 | 3 | 413 | |||
XM_011517591.2 | XP_011515893.1 | 3 | 386 | |||
XM_017013827.1 | XP_016869316.1 | 4 | 536 | |||
XM_017013828.1 | XP_016869317.1 | 2 | 815 | |||
GenBank mRNA | AK021829.1 | - | 2 | 141 | ||
BC025676.1 | - | 2 | 204 | 5203436 |