Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302652.1 | NP_001289581.1 | 2 | 229 | ||
NM_001302653.1 | NP_001289582.1 | 2 | 229 | |||
NM_001302654.1 | NP_001289583.1 | 2 | 229 | |||
NM_032374.4 | NP_115750.2 | 2 | 229 | |||
NR_126431.1 | 3 | 394 | ||||
NR_126432.1 | 2 | 229 | ||||
GenBank mRNA | AK223092.1 | - | 2 | 200 | ||
AK223103.1 | - | 2 | 200 | |||
BC007412.2 | - | 2 | 192 | 2989951 | ||
BF528062.1 | - | 3 | 297 | |||
BP317962.2 | - | 2 | 224 | |||
BP368962.1 | - | 2 | 223 | |||
BU181154.1 | - | 224 | ||||
BU956674.1 | - | 2 | 186 | |||
BX119699.1 | - | 2 | 109 | |||
BX340258.2 | - | 2 | 208 | |||
CB136383.1 | - | 2 | 224 | |||
CB993873.1 | - | 2 | 226 |