Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032423.2 | NP_115799.2 | 7 | 1533 | ||
XM_005259323.1 | XP_005259380.1 | 4 | 1116 | |||
XM_006723418.2 | XP_006723481.1 | 9 | 1781 | |||
XM_011527392.1 | XP_011525694.1 | 9 | 1777 | |||
XM_011527394.2 | XP_011525696.1 | 4 | 1649 | |||
XM_017027360.1 | XP_016882849.1 | 10 | 1898 | |||
XM_017027361.1 | XP_016882850.1 | 10 | 1894 | |||
XM_017027362.1 | XP_016882851.1 | 8 | 1693 | |||
XM_017027363.1 | XP_016882852.1 | 9 | 1687 | |||
XM_017027364.1 | XP_016882853.1 | 8 | 1642 | |||
XM_017027365.1 | XP_016882854.1 | 4 | 1192 | |||
GenBank mRNA | AB058730.1 | - | 1 | 630 | ||
AK056552.1 | - | 8 | 1765 | |||
AL832630.2 | - | 1 | 2356 | |||
AL833325.1 | - | 7 | 1533 | |||
BC101712.1 | - | 4 | 1149 | 8069218 | ||
BC111950.1 | - | 4 | 1161 | 8327418 |