Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM25 SIRNA Assays ›
Gene Symbol:
TMEM25
Gene Name:
transmembrane protein 25
Gene Aliases:
-
Chromosome Location:
Chr.11: 118531088 - 118546670 on Build GRCh38
UniGene ID:
Hs.564188
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS131342
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM25
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144034.1
NP_001137506.1
5
863
NM_001144035.1
NP_001137507.1
5
860
NM_001144036.1
NP_001137508.1
4
551
NM_001144037.1
NP_001137509.1
6
995
NM_001144038.1
NP_001137510.1
5
863
NM_001318755.1
NP_001305684.1
6
995
NM_001318757.1
NP_001305686.1
5
704
NM_032780.3
NP_116169.2
6
995
NR_134852.1
3
454
XM_005271703.1
XP_005271760.1
6
904
XM_006718926.2
XP_006718989.1
6
879
XM_006718927.2
XP_006718990.1
5
747
XM_011543037.2
XP_011541339.1
6
879
XM_011543038.2
XP_011541340.1
5
747
XM_017018422.1
XP_016873911.1
6
904
XM_017018423.1
XP_016873912.1
6
904
XM_017018424.1
XP_016873913.1
6
785
XM_017018425.1
XP_016873914.1
6
808
XM_017018426.1
XP_016873915.1
5
772
XM_017018427.1
XP_016873916.1
6
785
XM_017018428.1
XP_016873917.1
6
808
XM_017018429.1
XP_016873918.1
6
785
XM_017018430.1
XP_016873919.1
5
747
XM_017018431.1
XP_016873920.1
5
747
XM_017018432.1
XP_016873921.1
5
653
XM_017018433.1
XP_016873922.1
5
676
GenBank mRNA
AK027305.1
-
6
884
AK056325.1
-
4
437
AK096260.1
-
3
1567
AK124814.1
-
3
1713
AK225415.1
-
6
881
AK292359.1
-
5
747
AK293670.1
-
5
600
AY358919.1
-
5
748
BC042896.1
-
6
874
5168151
BC051841.1
-
4
731
5725777
DA760733.1
-
3
340
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline