Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024383.1 | NP_001019554.1 | 19, 20 | 4806 | ||
NM_014903.5 | NP_055718.4 | 19, 20 | 4806 | |||
XM_005269215.3 | XP_005269272.1 | 19, 20 | 8807 | |||
XM_011538940.2 | XP_011537242.1 | 18, 19 | 8766 | |||
XM_011538941.2 | XP_011537243.1 | 18, 19 | 8765 | |||
XM_011538944.2 | XP_011537246.1 | 17, 18 | 8274 | |||
XM_017020164.1 | XP_016875653.1 | 19, 20 | 8809 | |||
XM_017020165.1 | XP_016875654.1 | 18, 19 | 8767 | |||
XM_017020166.1 | XP_016875655.1 | 19, 20 | 4795 | |||
XM_017020167.1 | XP_016875656.1 | 18, 19 | 4726 | |||
XM_017020168.1 | XP_016875657.1 | 18, 19 | 8319 | |||
XM_017020169.1 | XP_016875658.1 | 17, 18 | 8276 | |||
XM_017020170.1 | XP_016875659.1 | 17, 18 | 8273 | |||
XM_017020171.1 | XP_016875660.1 | 11, 12 | 8102 | |||
XM_017020172.1 | XP_016875661.1 | 10, 11 | 3698 | |||
XM_017020173.1 | XP_016875662.1 | 10, 11 | 7611 | |||
XM_017020174.1 | XP_016875663.1 | 9, 10 | 7569 | |||
XM_017020175.1 | XP_016875664.1 | 10, 11 | 3490 | |||
GenBank mRNA | AB023155.2 | - | 4, 5 | 1316 | ||
AF397731.1 | - | 18, 19 | 4390 | |||
AJ488201.1 | - | 19, 20 | 4678 | |||
AK298525.1 | - | 3, 4 | 520 |