Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033117.3 | NP_149108.1 | 2 | 234 | ||
NR_027125.1 | 2 | 234 | ||||
NR_027126.1 | 2 | 234 | ||||
XM_006717319.1 | XP_006717382.1 | 2 | 190 | |||
GenBank mRNA | AK057676.1 | - | 2 | 210 | ||
AK299625.1 | - | 2 | 210 | |||
AK301050.1 | - | 2 | 190 | |||
AK310008.1 | - | 2 | 191 | |||
AK310807.1 | - | 2 | 176 | |||
AL389986.1 | - | 2 | 162 | |||
BC008942.2 | - | 2 | 181 | 2822634 | ||
BC019319.1 | - | 2 | 181 | 2822634 | ||
BP870502.1 | - | 2 | 213 | |||
DA308339.1 | - | 2 | 234 |