Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286250.1 | NP_001273179.1 | 4, 5 | 769 | ||
NM_001286252.1 | NP_001273181.1 | 5, 6 | 877 | |||
NM_001286254.2 | NP_001273183.1 | 4, 5 | 562 | |||
NM_001323252.1 | NP_001310181.1 | 6, 7 | 924 | |||
NM_001323253.1 | NP_001310182.1 | 5, 6 | 877 | |||
NM_001323256.1 | NP_001310185.1 | 5, 6 | 877 | |||
NM_001323258.1 | NP_001310187.1 | 6, 7 | 2621 | |||
NM_001323259.1 | NP_001310188.1 | 5, 6 | 877 | |||
NM_001323260.1 | NP_001310189.1 | 5, 6 | 877 | |||
NM_001323261.1 | NP_001310190.1 | 3, 4 | 391 | |||
NM_001323262.1 | NP_001310191.1 | 3, 4 | 391 | |||
NM_001323263.1 | NP_001310192.1 | 3, 4 | 391 | |||
NM_052904.3 | NP_443136.2 | 5, 6 | 877 | |||
NR_136579.1 | 3, 4 | 391 | ||||
NR_136580.1 | 5, 6 | 877 | ||||
XM_005266813.4 | XP_005266870.1 | 3, 4 | 282 | |||
XM_017010229.1 | XP_016865718.1 | 2, 3 | 160 | |||
GenBank mRNA | AB067487.1 | - | 3, 4 | 291 | ||
AK055292.1 | - | 5, 6 | 763 | |||
AK295479.1 | - | 4, 5 | 594 | |||
BC047491.1 | - | 4, 5 | 442 | |||
BC071627.1 | - | 5, 6 | 752 | 5245540 | ||
BG181952.1 | - | 1, 2 | 93 | |||
BI601157.1 | - | 4, 5 | 447 | |||
BX645412.1 | - | 3, 4 | 398 | |||
DA255882.1 | - | 1, 2 | 23 |