Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOSTRIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039724.3 | NP_001034813.2 | 14 | 1333 | ||
NM_001171631.1 | NP_001165102.1 | 19 | 2205 | |||
NM_001171632.1 | NP_001165103.1 | 13 | 1249 | |||
NM_052946.3 | NP_443178.2 | 13 | 1240 | |||
XM_005246270.1 | XP_005246327.1 | 14 | 1392 | |||
XM_006712233.3 | XP_006712296.1 | 15 | 1455 | |||
XM_011510540.2 | XP_011508842.1 | 15 | 1455 | |||
XM_011510543.2 | XP_011508845.1 | 8 | 908 | |||
XM_017003278.1 | XP_016858767.1 | 14 | 1284 | |||
XM_017003279.1 | XP_016858768.1 | 14 | 1778 | |||
XM_017003280.1 | XP_016858769.1 | 13 | 1255 | |||
GenBank mRNA | AJ532842.1 | - | 14 | 1276 | ||
AK002203.2 | - | 14 | 1305 | |||
AK093444.1 | - | 19 | 2203 | |||
AK123698.1 | - | 3 | 1317 | |||
AK290254.1 | - | 14 | 1299 | |||
BC014189.2 | - | 13 | 1240 | 4650302 | ||
BC093072.1 | - | 14 | 1280 | 30327154 | ||
BF114987.1 | - | 3 | 342 | |||
BQ899074.1 | - | 1 | 22 | |||
DQ402497.1 | - | 13 | 1192 |