Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289112.1 | NP_001276041.1 | 3 | 528 | ||
NM_130808.2 | NP_570720.1 | 3 | 1156 | |||
NM_153429.1 | NP_702907.1 | 4 | 516 | |||
XM_011512406.2 | XP_011510708.1 | 2 | 303 | |||
XM_011512407.2 | XP_011510709.1 | 2 | 560 | |||
XM_011512408.2 | XP_011510710.1 | 2 | 1688 | |||
XM_017005693.1 | XP_016861182.1 | 6 | 1909 | |||
XM_017005694.1 | XP_016861183.1 | 2 | 280 | |||
XM_017005695.1 | XP_016861184.1 | 3 | 549 | |||
GenBank mRNA | AF465770.1 | - | 6 | 856 | ||
AF465771.1 | - | 4 | 516 | |||
AK056175.1 | - | 3 | 628 | |||
AK128117.1 | - | 7 | 2406 | |||
AK295557.1 | - | 3 | 443 | |||
BC014396.1 | - | 3 | 714 | 3162811 | ||
BC075046.2 | - | 3 | 305 | 30915576 | ||
DC330179.1 | - | 3 | 528 |