Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282855.1 | NP_001269784.1 | 8 | 1958 | ||
NM_001321293.1 | NP_001308222.1 | 8 | 1970 | |||
NM_178827.4 | NP_849149.3 | 8 | 1558 | |||
NR_104244.1 | 8 | 1545 | ||||
XM_005250161.3 | XP_005250218.1 | 9 | 1673 | |||
XM_005250162.4 | XP_005250219.1 | 8 | 1539 | |||
XM_011515833.2 | XP_011514135.1 | 9 | 1486 | |||
XM_011515834.2 | XP_011514136.1 | 8 | 1791 | |||
XM_017011771.1 | XP_016867260.1 | 9 | 1472 | |||
XM_017011772.1 | XP_016867261.1 | 3 | 1134 | |||
GenBank mRNA | AK093153.1 | - | 8 | 1478 | ||
BC026173.1 | - | 8 | 1549 | 4838069 | ||
BC032838.2 | - | 8 | 1483 | |||
BC036121.1 | - | 8 | 1958 | 5296826 | ||
BC068607.1 | - | 8 | 1483 | |||
BC091520.1 | - | 8 | 1413 | 30379054 | ||
BC128181.1 | - | 8 | 1457 | 40113310 |