Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM122B SIRNA Assays ›
          Gene Symbol:
          FAM122B
          Gene Name:
          family with sequence similarity 122B
          Gene Aliases:
          SPACIA2
          Chromosome Location:
          Chr.X: 134769566 - 134797232 on Build GRCh38
          UniGene ID:
          Hs.404706
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS136155
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM122B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166599.2 NP_001160071.1 8 1492
          NM_001166600.2 NP_001160072.1 10 861
          NM_001170756.1 NP_001164227.1 9 749
          NM_001170757.1 NP_001164228.1 9 580
          NM_145284.5 NP_660327.2 8 695
          XM_006724730.2 XP_006724793.1 9 1608
          XM_006724732.2 XP_006724795.1 9 1608
          XM_006724733.3 XP_006724796.1 8 1545
          XM_011531282.1 XP_011529584.1 9 1620
          XM_011531283.1 XP_011529585.1 9 1617
          XM_011531284.1 XP_011529586.1 9 1608
          XM_011531285.1 XP_011529587.1 9 1605
          XM_011531286.1 XP_011529588.1 8 1563
          XM_011531287.1 XP_011529589.1 9 1620
          XM_011531288.1 XP_011529590.1 8 1545
          XM_011531289.1 XP_011529591.1 9 1617
          XM_011531290.1 XP_011529592.1 8 1548
          XM_011531291.1 XP_011529593.1 9 1620
          XM_011531292.1 XP_011529594.1 9 1617
          XM_011531293.1 XP_011529595.1 9 1605
          XM_011531294.1 XP_011529596.1 8 1548
          XM_017029306.1 XP_016884795.1 8 1563
          XM_017029307.1 XP_016884796.1 8 1560
          GenBank mRNA AB541014.1 - 9 602
          AB541015.1 - 9 599
          AK124650.1 - 10 834
          AK124940.1 - 8 481
          AK125991.1 - 9 652
          AK292517.1 - 8 614
          AK297720.1 - 9 580
          AL832218.1 - 8 625
          BC019221.2 - 8 1471 3841054
          BC032419.2 - 4 268
          BC110846.1 - 8 1528 6046376
          BX538218.1 - 9 972

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0