Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290083.1 | NP_001277012.1 | 5 | 624 | ||
NM_001290084.1 | NP_001277013.1 | 4 | 458 | |||
NM_001290085.1 | NP_001277014.1 | 4 | 465 | |||
NM_145295.3 | NP_660338.1 | 4 | 469 | |||
XM_011527780.2 | XP_011526082.1 | 4 | 357 | |||
GenBank mRNA | AK074846.1 | - | 4 | 377 | ||
AK095639.1 | - | 4 | 395 | |||
BC006279.2 | - | 4 | 390 | |||
BC069232.1 | - | 4 | 354 | 6645413 | ||
BC098416.1 | - | 4 | 440 | |||
DA786363.1 | - | 4 | 425 | |||
DB043358.1 | - | 4 | 469 |