Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BEND7 SIRNA Assays ›
Gene Symbol:
BEND7
Gene Name:
BEN domain containing 7
Gene Aliases:
C10orf30
Chromosome Location:
Chr.10: 13438484 - 13528772 on Build GRCh38
UniGene ID:
Hs.498740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS137613
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BEND7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100912.1
NP_001094382.1
4
812
NM_152751.2
NP_689964.2
5
943
XM_005252402.4
XP_005252459.1
5
1041
XM_011519390.2
XP_011517692.1
4
734
XM_011519391.2
XP_011517693.1
4
734
XM_011519392.2
XP_011517694.1
5
1040
XM_011519393.2
XP_011517695.1
4
829
XM_011519394.2
XP_011517696.1
6
976
XM_011519395.2
XP_011517697.1
4
1336
XM_011519396.2
XP_011517698.1
5
1427
XM_011519397.2
XP_011517699.1
5
982
XM_011519398.2
XP_011517700.1
4
734
XM_011519399.2
XP_011517701.1
4
734
XM_011519400.2
XP_011517702.1
4
734
XM_011519401.2
XP_011517703.1
4
734
XM_011519402.2
XP_011517704.1
4
734
XM_011519403.2
XP_011517705.1
4
734
XM_011519404.2
XP_011517706.1
4
734
XM_011519405.2
XP_011517707.1
4
734
XM_011519406.2
XP_011517708.1
4
734
XM_011519407.1
XP_011517709.1
4
734
XM_011519408.2
XP_011517710.1
4
734
XM_011519410.2
XP_011517712.1
4
734
XM_011519411.2
XP_011517713.1
4
734
XM_011519412.1
XP_011517714.1
4
734
XM_011519413.1
XP_011517715.1
4
734
XM_011519414.1
XP_011517716.1
4
734
XM_017015915.1
XP_016871404.1
6
961
XM_017015916.1
XP_016871405.1
4
734
XM_017015917.1
XP_016871406.1
4
734
XM_017015918.1
XP_016871407.1
5
975
XM_017015919.1
XP_016871408.1
5
997
GenBank mRNA
AK097602.1
-
5
943
BC031618.1
-
4
781
5172600
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline