Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZBTB8OS SIRNA Assays ›
          Gene Symbol:
          ZBTB8OS
          Gene Name:
          zinc finger and BTB domain containing 8 opposite strand
          Gene Aliases:
          ARCH, ARCH2
          Chromosome Location:
          Chr.1: 32620820 - 32651013 on Build GRCh38
          UniGene ID:
          Hs.655921
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS139097
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZBTB8OS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001308135.1 NP_001295064.1 2, 3 289
          NM_001308136.1 NP_001295065.1 3, 4 310
          NM_001308137.1 NP_001295066.1 2, 3 289
          NM_001308138.1 NP_001295067.1 2, 3 354
          NM_001308139.1 NP_001295068.1 3, 4 348
          NM_001308140.1 NP_001295069.1 3, 4 341
          NM_001308141.1 NP_001295070.1 3, 4 314
          NM_178547.3 NP_848642.1 3, 4 310
          XM_011541327.2 XP_011539629.1 3, 4 733
          XM_011541328.1 XP_011539630.1 3, 4 335
          XM_011541329.2 XP_011539631.1 3, 4 235
          XM_011541330.2 XP_011539632.1 2, 3 721
          XM_011541334.2 XP_011539636.1 2, 3 680
          XM_011541335.2 XP_011539637.1 3, 4 287
          XM_011541337.2 XP_011539639.1 3, 4 322
          XM_011541338.2 XP_011539640.1 2, 3 681
          XM_017001134.1 XP_016856623.1 3, 4 321
          XM_017001135.1 XP_016856624.1 3, 4 733
          XM_017001137.1 XP_016856626.1 3, 4 169
          XM_017001138.1 XP_016856627.1 2, 3 207
          GenBank mRNA AA316493.1 - 3, 4 245
          AK289480.1 - 3, 4 652
          AY099494.1 - 3, 4 210
          AY151084.1 - 3, 4 304
          AY151085.1 - 3, 4 304
          BC058843.1 - 3, 4 280 6745279
          BE780045.1 - 2, 3 251
          BF216169.1 - 3, 4 285
          BG059414.1 - 2, 3 225
          BQ187597.1 - 3, 4 253
          CK819207.1 - 3, 4 245
          CN344731.1 - 3, 4 250
          DT220608.1 - 2, 3 314

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA splicing, via endonucleolytic cleavage and ligation
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0