Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004313.1 | NP_001004313.1 | 4, 5 | 775 | ||
XM_005256634.4 | XP_005256691.1 | 3, 4 | 710 | |||
XM_017024616.1 | XP_016880105.1 | 4, 5 | 319 | |||
XM_017024617.1 | XP_016880106.1 | 3, 4 | 711 | |||
GenBank mRNA | AK289624.1 | - | 4, 5 | 431 | ||
AK295928.1 | - | 3, 4 | 440 | |||
AY550194.1 | - | 4, 5 | 775 | |||
BC127705.1 | - | 4, 5 | 339 | 40127270 | ||
BC136838.1 | - | 4, 5 | 341 | 9020829 | ||
BC136840.1 | - | 4, 5 | 341 | 9020831 | ||
EF126147.1 | - | 3, 4 | 267 |