Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCSER1 SIRNA Assays ›
          Gene Symbol:
          CCSER1
          Gene Name:
          coiled-coil serine rich protein 1
          Gene Aliases:
          FAM190A
          Chromosome Location:
          Chr.4: 90127394 - 91602219 on Build GRCh38
          UniGene ID:
          Hs.654735
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS140425
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCSER1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145065.1 NP_001138537.1 2 1331
          NM_207491.2 NP_997374.1 2 1345
          XM_011531936.1 XP_011530238.1 2 1470
          XM_011531937.2 XP_011530239.1 2 1871
          XM_011531938.2 XP_011530240.1 2 1195
          XM_011531939.2 XP_011530241.1 2 2695
          XM_011531941.2 XP_011530243.1 2 1470
          XM_011531942.2 XP_011530244.1 2 1470
          XM_011531943.2 XP_011530245.1 2 1470
          XM_011531945.1 XP_011530247.1 2 1470
          XM_011531946.2 XP_011530248.1 2 1470
          XM_011531947.2 XP_011530249.1 2 1470
          XM_011531948.1 XP_011530250.1 2 1470
          XM_011531949.2 XP_011530251.1 2 1470
          XM_011531950.2 XP_011530252.1 2 1470
          XM_011531951.2 XP_011530253.1 2 1470
          XM_011531955.2 XP_011530257.1 2 1470
          XM_011531956.2 XP_011530258.1 2 1470
          XM_011531957.2 XP_011530259.1 2 1470
          XM_011531958.2 XP_011530260.1 2 1470
          XM_017008194.1 XP_016863683.1 2 1193
          XM_017008195.1 XP_016863684.1 2 1470
          XM_017008196.1 XP_016863685.1 2 1470
          XM_017008197.1 XP_016863686.1 2 1470
          XM_017008198.1 XP_016863687.1 2 1470
          XM_017008199.1 XP_016863688.1 2 1470
          XM_017008200.1 XP_016863689.1 2 1470
          XM_017008201.1 XP_016863690.1 2 1470
          XM_017008202.1 XP_016863691.1 2 1470
          GenBank mRNA AB051467.1 - 2 1279
          AK097436.1 - 2 1331
          AK310825.1 - 2 1329
          BC051694.1 - 2 1345 5296943

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0