Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026781.1 | 2 | 1046 | |||
GenBank mRNA | AK056013.1 | - | 2 | 1046 | ||
BC119778.1 | - | 1 | 729 | |||
BC122871.1 | - | 1 | 730 |
Gene Symbol: LINC00574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026780.1 | 1 | 86 | |||
GenBank mRNA | DA918406.1 | - | 1 | 86 |