Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGFR1 SIRNA Assays ›
Gene Symbol:
FGFR1
Gene Name:
fibroblast growth factor receptor 1
Gene Aliases:
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Chromosome Location:
Chr.8: 38411138 - 38468834 on Build GRCh38
UniGene ID:
Hs.264887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS142013
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGFR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174063.1
NP_001167534.1
6
1583
NM_001174064.1
NP_001167535.1
7
1651
NM_001174065.1
NP_001167536.1
6
897
NM_001174066.1
NP_001167537.1
5
636
NM_001174067.1
NP_001167538.1
7
1057
NM_015850.3
NP_056934.2
6
1577
NM_023105.2
NP_075593.1
5
1316
NM_023106.2
NP_075594.1
5
1310
NM_023110.2
NP_075598.2
6
1583
XM_006716303.2
XP_006716366.1
6
1384
XM_006716304.1
XP_006716367.1
6
900
XM_006716306.2
XP_006716369.1
6
1378
XM_006716307.1
XP_006716370.1
6
894
XM_006716309.3
XP_006716372.1
6
1354
XM_006716310.2
XP_006716373.1
5
1117
XM_006716311.1
XP_006716374.1
5
515
XM_006716312.1
XP_006716375.1
5
585
XM_006716313.2
XP_006716376.1
5
1111
XM_006716314.1
XP_006716377.1
5
589
XM_011544443.2
XP_011542745.1
6
889
XM_011544444.1
XP_011542746.1
6
878
XM_011544445.2
XP_011542747.1
6
897
XM_011544446.2
XP_011542748.1
6
884
XM_011544447.2
XP_011542749.1
6
884
XM_011544448.1
XP_011542750.1
5
488
XM_011544449.1
XP_011542751.1
5
482
XM_011544450.2
XP_011542752.1
5
482
XM_011544451.1
XP_011542753.1
4
351
XM_011544452.2
XP_011542754.1
6
972
XM_017013219.1
XP_016868708.1
6
885
XM_017013220.1
XP_016868709.1
6
878
XM_017013221.1
XP_016868710.1
7
1093
XM_017013222.1
XP_016868711.1
6
1384
XM_017013223.1
XP_016868712.1
6
894
XM_017013224.1
XP_016868713.1
6
1378
XM_017013225.1
XP_016868714.1
6
861
XM_017013226.1
XP_016868715.1
5
488
XM_017013227.1
XP_016868716.1
5
482
XM_017013228.1
XP_016868717.1
5
1111
XM_017013230.1
XP_016868719.1
4
442
XM_017013231.1
XP_016868720.1
6
982
GenBank mRNA
AB208919.1
-
7
1438
AK222718.1
-
6
1376
AK223126.1
-
5
1111
AK291754.1
-
6
776
AK292470.1
-
5
636
AK309947.1
-
7
936
BC015035.1
-
7
1311
3911101
BC091494.1
-
4
447
30387634
BX649167.1
-
2
2925
EU826596.1
-
4
368
EU826597.1
-
4
374
FJ809916.1
-
6
1586
FJ809917.1
-
6
1586
KU178066.1
-
5
641
KU178067.1
-
4
374
KU178068.1
-
4
368
KU178069.1
-
4
374
KU178070.1
-
4
368
M34185.1
-
5
629
M34186.1
-
5
894
M34187.1
-
5
790
M34188.1
-
5
890
M34641.1
-
5
644
M37722.1
-
5
596
M60485.1
-
7
1306
M63887.1
-
5
641
M63888.1
-
5
641
M63889.1
-
5
518
X51803.1
-
6
758
X52833.1
-
5
728
X57118.1
-
6
779
X57119.1
-
5
536
X57120.1
-
6
797
X57121.1
-
6
803
X57122.1
-
5
530
X66945.1
-
6
1367
Y00665.1
-
2
41
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
skeletal system development
angiogenesis
ureteric bud development
in utero embryonic development
organ induction
neuron migration
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
transcription, DNA-templated
protein phosphorylation
sensory perception of sound
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of phospholipase activity
positive regulation of phospholipase C activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
stem cell population maintenance
motogenic signaling involved in postnatal olfactory bulb interneuron migration
ventricular zone neuroblast division
embryonic limb morphogenesis
midbrain development
neuron projection development
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
chordate embryonic development
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cell differentiation
positive regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of cell cycle
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
paraxial mesoderm development
regulation of lateral mesodermal cell fate specification
cell maturation
skeletal system morphogenesis
mesenchymal cell differentiation
regulation of sensory perception of pain
positive regulation of cardiac muscle cell proliferation
auditory receptor cell development
branching involved in salivary gland morphogenesis
lung-associated mesenchyme development
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
regulation of stem cell proliferation
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
regulation of extrinsic apoptotic signaling pathway in absence of ligand
glycoprotein binding
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein complex binding
identical protein binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix