Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPH3 SIRNA Assays ›
          Gene Symbol:
          HNRNPH3
          Gene Name:
          heterogeneous nuclear ribonucleoprotein H3
          Gene Aliases:
          2H9, HNRPH3
          Chromosome Location:
          Chr.10: 68331174 - 68343196 on Build GRCh38
          UniGene ID:
          Hs.643472
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS142482
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HNRNPH3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001322434.1 NP_001309363.1 8, 9 1108
          NM_001322436.1 NP_001309365.1 8, 9 1070
          NM_001322437.1 NP_001309366.1 8, 9 1173
          NM_001322438.1 NP_001309367.1 8, 9 1000
          NM_001322439.1 NP_001309368.1 8, 9 1063
          NM_001322440.1 NP_001309369.1 8, 9 1025
          NM_001322441.1 NP_001309370.1 8, 9 1128
          NM_001322442.1 NP_001309371.1 8, 9 1090
          NM_001322443.1 NP_001309372.1 8, 9 1071
          NM_001322444.1 NP_001309373.1 6, 7 765
          NM_001322445.1 NP_001309374.1 7, 8 906
          NM_001322446.1 NP_001309375.1 7, 8 950
          NM_001322447.1 NP_001309376.1 7, 8 931
          NM_001322448.1 NP_001309377.1 7, 8 969
          NM_001322449.1 NP_001309378.1 7, 8 1034
          NM_001322450.1 NP_001309379.1 7, 8 951
          NM_001322451.1 NP_001309380.1 7, 8 886
          NM_001322452.1 NP_001309381.1 7, 8 905
          NM_001322453.1 NP_001309382.1 7, 8 924
          NM_012207.2 NP_036339.1 8, 9 1089
          NM_021644.3 NP_067676.2 8, 9 1044
          XM_011539742.1 XP_011538044.1 6, 7 736
          XM_017016168.1 XP_016871657.1 8, 9 1519
          XM_017016169.1 XP_016871658.1 7, 8 1380
          GenBank mRNA AF132361.1 - 8, 9 933
          AF132362.1 - 6, 7 990
          AK091411.1 - 6, 7 2961
          AK223441.1 - 8, 9 1041
          AK291547.1 - 8, 9 996
          AK295316.1 - 6, 7 680
          AL117395.1 - 5, 6 2311
          BC004511.2 - 5, 6 445
          BC039824.1 - 8, 9 980 6061550

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          RNA processing
          RNA splicing
          epithelial cell differentiation
          nucleotide binding
          RNA binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
          git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix