Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018953.3 | NP_061826.1 | 1 | 157 | ||
GenBank mRNA | BM786543.1 | - | 1 | 69 |