Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136007.1 | NP_001129479.1 | 7, 8 | 538 | ||
NM_001136008.1 | NP_001129480.1 | 5, 6 | 332 | |||
NM_001136011.1 | NP_001129483.1 | 6, 7 | 416 | |||
NM_001136012.1 | NP_001129484.1 | 5, 6 | 332 | |||
NM_005971.3 | NP_005962.1 | 5, 6 | 332 | |||
NM_021910.2 | NP_068710.1 | 6, 7 | 416 | |||
XM_005258996.4 | XP_005259053.1 | 4, 5 | 237 | |||
XM_011527032.2 | XP_011525334.1 | 7, 8 | 545 | |||
XM_017026877.1 | XP_016882366.1 | 5, 6 | 282 | |||
XM_017026878.1 | XP_016882367.1 | 4, 5 | 202 | |||
GenBank mRNA | BC005238.1 | - | 4, 5 | 146 | 3997821 | |
BC090044.1 | - | 5, 6 | 281 | 30395606 | ||
BE793135.1 | - | 3, 4 | 106 | |||
BI715233.1 | - | 4, 5 | 122 | |||
BM549935.1 | - | 3, 4 | 144 | |||
BQ676323.1 | - | 6, 7 | 364 | |||
BQ682674.1 | - | 6, 7 | 662 | |||
BT006712.1 | - | 3, 4 | 90 | |||
BU185141.1 | - | 5, 6 | 219 | |||
CA948888.1 | - | 2, 3 | 59 | |||
CB128589.1 | - | 4, 5 | 145 | |||
CR456945.1 | - | 3, 4 | 90 | |||
CR542197.1 | - | 3, 4 | 90 | |||
DA890604.1 | - | 4, 5 | 174 | |||
U28249.1 | - | 6, 7 | 349 | |||
X93036.1 | - | 4, 5 | 149 |