Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POU5F1P4 SIRNA Assays ›
          Gene Symbol:
          POU5F1P4 POU5F1B POU5F1P3
          Gene Name:
          POU class 5 homeobox 1 pseudogene 4
          POU class 5 homeobox 1B
          POU class 5 homeobox 1 pseudogene 3
          Gene Aliases:
          Oct4-pg4, POU5FLC1
          OCT4-PG1, OCT4PG1, OTF3C, OTF3P1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8
          OCT4-pg3, OTF3L, POU5F1L, POU5FLC12
          Chromosome Location:
          Chr.1: 155433180 - 155434262 on Build GRCh38
          Chromosome Location:
          Chr.8: 127415612 - 127417210 on Build GRCh38
          Chromosome Location:
          Chr.12: 8133769 - 8134852 on Build GRCh38
          UniGene ID:
          Hs.632482
          Hs.450254
          Hs.646545
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID HSS143404
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POU5F1P4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA GU480890.1 - 1 281
          HQ122798.1 - 1 282
          HQ122800.1 - 1 282
          HQ122804.1 - 1 282
          HQ122807.1 - 1 282
          HQ122811.1 - 1 282
          HQ122858.1 - 1 282
          Gene Symbol: POU5F1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001159542.1 NP_001153014.1 1 535
          GenBank mRNA AF268615.1 - 1 280
          AF268618.1 - 1 280
          DQ486513.1 - 1 535
          GU480835.1 - 1 281
          GU480840.1 - 1 281
          GU480841.1 - 1 281
          GU480842.1 - 1 281
          GU480844.1 - 1 281
          GU480845.1 - 1 281
          GU480849.1 - 1 282
          GU480850.1 - 1 282
          GU480851.1 - 1 282
          GU480864.1 - 1 281
          GU480868.1 - 1 281
          GU480878.1 - 1 281
          GU480880.1 - 1 281
          GU480883.1 - 1 281
          GU480884.1 - 1 281
          GU480885.1 - 1 282
          GU480886.1 - 1 281
          GU480889.1 - 1 281
          GU480896.1 - 1 281
          GU480899.1 - 1 282
          GU480902.1 - 1 281
          GU480905.1 - 1 281
          GU480908.1 - 1 281
          GU480910.1 - 1 283
          GU480911.1 - 1 281
          GU480912.1 - 1 281
          GU480913.1 - 1 281
          HQ122710.1 - 1 281
          HQ122711.1 - 1 281
          HQ122712.1 - 1 281
          HQ122713.1 - 1 281
          HQ122714.1 - 1 281
          HQ122715.1 - 1 281
          HQ122716.1 - 1 281
          HQ122717.1 - 1 281
          HQ122718.1 - 1 281
          HQ122719.1 - 1 281
          HQ122720.1 - 1 281
          HQ122721.1 - 1 281
          HQ122750.1 - 1 281
          HQ122751.1 - 1 281
          HQ122752.1 - 1 281
          HQ122781.1 - 1 281
          HQ122782.1 - 1 281
          HQ122783.1 - 1 281
          HQ122784.1 - 1 281
          HQ122785.1 - 1 281
          HQ122786.1 - 1 281
          HQ122787.1 - 1 281
          HQ122788.1 - 1 281
          HQ122789.1 - 1 281
          HQ122790.1 - 1 281
          HQ122816.1 - 1 281
          HQ122817.1 - 1 281
          HQ122818.1 - 1 281
          HQ122819.1 - 1 281
          HQ122820.1 - 1 281
          HQ122821.1 - 1 281
          HQ122822.1 - 1 281
          HQ122823.1 - 1 281
          HQ122824.1 - 1 281
          HQ122825.1 - 1 281
          HQ122826.1 - 1 281
          HQ122841.1 - 1 281
          HQ122842.1 - 1 281
          HQ122843.1 - 1 281
          HQ122844.1 - 1 281
          HQ122845.1 - 1 281
          HQ122846.1 - 1 281
          HQ122847.1 - 1 281
          HQ122848.1 - 1 281
          HQ122849.1 - 1 281
          HQ122850.1 - 1 282
          HQ122851.1 - 1 281
          HQ122852.1 - 1 281
          HQ122853.1 - 1 281
          HQ122854.1 - 1 281
          HQ122855.1 - 1 281
          HQ122856.1 - 1 281
          HQ122857.1 - 1 281
          HQ122863.1 - 1 281
          HQ122864.1 - 1 281
          HQ122865.1 - 1 281
          HQ122866.1 - 1 281
          HQ122867.1 - 1 281
          HQ122868.1 - 1 281
          HQ122869.1 - 1 281
          HQ122870.1 - 1 281
          HQ122871.1 - 1 281
          HQ122872.1 - 1 281
          HQ122873.1 - 1 281
          HQ122874.1 - 1 281
          HQ122875.1 - 1 281
          HQ122876.1 - 1 281
          HQ122877.1 - 1 281
          HQ122878.1 - 1 281
          HQ122879.1 - 1 281
          HQ122880.1 - 1 281
          HQ122881.1 - 1 281
          HQ122882.1 - 1 281
          HQ122883.1 - 1 281
          HQ122884.1 - 1 281
          HQ122885.1 - 1 281
          HQ122886.1 - 1 281
          HQ122887.1 - 1 281
          HQ122891.1 - 1 281
          HQ122892.1 - 1 281
          HQ122893.1 - 1 281
          HQ122894.1 - 1 280
          HQ122895.1 - 1 281
          HQ122896.1 - 1 281
          HQ122897.1 - 1 281
          HQ122898.1 - 1 281
          HQ122899.1 - 1 281
          HQ122900.1 - 1 281
          HQ122901.1 - 1 281
          HQ122902.1 - 1 281
          HQ122903.1 - 1 281
          Gene Symbol: POU5F1P3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA GU480888.1 - 1 282

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc