Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POU5F1P4 SIRNA Assays ›
          Gene Symbol:
          POU5F1P4 POU5F1B POU5F1P3
          Gene Name:
          POU class 5 homeobox 1 pseudogene 4
          POU class 5 homeobox 1B
          POU class 5 homeobox 1 pseudogene 3
          Gene Aliases:
          Oct4-pg4, POU5FLC1
          OCT4-PG1, OCT4PG1, OTF3C, OTF3P1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8
          OCT4-pg3, OTF3L, POU5F1L, POU5FLC12
          Chromosome Location:
          Chr.1: 155433180 - 155434262 on Build GRCh38
          Chromosome Location:
          Chr.8: 127415612 - 127417210 on Build GRCh38
          Chromosome Location:
          Chr.12: 8133769 - 8134852 on Build GRCh38
          UniGene ID:
          Hs.632482
          Hs.450254
          Hs.646545
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID HSS143405
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POU5F1P4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_034180.1 1 530
          GenBank mRNA AF268613.1 - 1 530
          DQ851566.1 - 1 530
          GU480814.1 - 1 531
          GU480831.1 - 1 532
          GU480833.1 - 1 531
          GU480834.1 - 1 531
          GU480836.1 - 1 531
          GU480839.1 - 1 531
          GU480848.1 - 1 531
          GU480852.1 - 1 531
          GU480853.1 - 1 532
          GU480854.1 - 1 531
          GU480855.1 - 1 529
          GU480856.1 - 1 531
          GU480857.1 - 1 531
          GU480858.1 - 1 531
          GU480860.1 - 1 530
          GU480861.1 - 1 532
          GU480862.1 - 1 531
          GU480863.1 - 1 531
          GU480865.1 - 1 532
          GU480866.1 - 1 531
          GU480867.1 - 1 531
          GU480869.1 - 1 531
          GU480870.1 - 1 531
          GU480871.1 - 1 531
          GU480872.1 - 1 531
          GU480873.1 - 1 531
          GU480875.1 - 1 531
          GU480879.1 - 1 531
          GU480881.1 - 1 531
          GU480882.1 - 1 531
          GU480890.1 - 1 530
          GU480894.1 - 1 531
          GU480895.1 - 1 531
          GU480898.1 - 1 530
          GU480900.1 - 1 530
          GU480904.1 - 1 531
          GU480914.1 - 1 530
          HQ122676.1 - 1 531
          HQ122677.1 - 1 531
          HQ122678.1 - 1 529
          HQ122679.1 - 1 530
          HQ122680.1 - 1 531
          HQ122681.1 - 1 531
          HQ122683.1 - 1 532
          HQ122684.1 - 1 531
          HQ122685.1 - 1 529
          HQ122686.1 - 1 531
          HQ122687.1 - 1 531
          HQ122689.1 - 1 529
          HQ122690.1 - 1 531
          HQ122691.1 - 1 531
          HQ122695.1 - 1 530
          HQ122696.1 - 1 530
          HQ122697.1 - 1 530
          HQ122698.1 - 1 531
          HQ122700.1 - 1 531
          HQ122701.1 - 1 531
          HQ122703.1 - 1 531
          HQ122704.1 - 1 531
          HQ122705.1 - 1 531
          HQ122707.1 - 1 530
          HQ122708.1 - 1 531
          HQ122709.1 - 1 531
          HQ122731.1 - 1 531
          HQ122732.1 - 1 531
          HQ122733.1 - 1 531
          HQ122734.1 - 1 531
          HQ122735.1 - 1 531
          HQ122736.1 - 1 531
          HQ122737.1 - 1 531
          HQ122738.1 - 1 531
          HQ122739.1 - 1 531
          HQ122740.1 - 1 531
          HQ122741.1 - 1 531
          HQ122742.1 - 1 531
          HQ122743.1 - 1 531
          HQ122744.1 - 1 531
          HQ122745.1 - 1 531
          HQ122746.1 - 1 531
          HQ122747.1 - 1 531
          HQ122748.1 - 1 531
          HQ122756.1 - 1 531
          HQ122757.1 - 1 531
          HQ122758.1 - 1 531
          HQ122759.1 - 1 531
          HQ122760.1 - 1 531
          HQ122762.1 - 1 531
          HQ122764.1 - 1 531
          HQ122765.1 - 1 531
          HQ122766.1 - 1 531
          HQ122767.1 - 1 531
          HQ122769.1 - 1 530
          HQ122770.1 - 1 531
          HQ122772.1 - 1 531
          HQ122773.1 - 1 531
          HQ122774.1 - 1 531
          HQ122775.1 - 1 531
          HQ122776.1 - 1 531
          HQ122777.1 - 1 530
          HQ122779.1 - 1 531
          HQ122780.1 - 1 531
          HQ122798.1 - 1 531
          HQ122799.1 - 1 530
          HQ122800.1 - 1 532
          HQ122801.1 - 1 531
          HQ122802.1 - 1 529
          HQ122803.1 - 1 532
          HQ122804.1 - 1 531
          HQ122805.1 - 1 531
          HQ122806.1 - 1 531
          HQ122807.1 - 1 531
          HQ122808.1 - 1 531
          HQ122809.1 - 1 529
          HQ122810.1 - 1 531
          HQ122811.1 - 1 531
          HQ122812.1 - 1 531
          HQ122813.1 - 1 531
          HQ122814.1 - 1 529
          HQ122815.1 - 1 531
          HQ122832.1 - 1 531
          HQ122833.1 - 1 531
          HQ122834.1 - 1 531
          HQ122835.1 - 1 529
          HQ122836.1 - 1 531
          HQ122837.1 - 1 531
          HQ122838.1 - 1 529
          HQ122839.1 - 1 531
          HQ122840.1 - 1 531
          HQ122858.1 - 1 531
          HQ122860.1 - 1 531
          HQ122862.1 - 1 531
          HQ122888.1 - 1 531
          HQ122889.1 - 1 531
          HQ122890.1 - 1 531
          HQ122904.1 - 1 531
          HQ122905.1 - 1 531
          HQ122907.1 - 1 531
          HQ122908.1 - 1 531
          HQ122909.1 - 1 532
          HQ122910.1 - 1 531
          HQ122911.1 - 1 531
          HQ122912.1 - 1 531
          HQ122913.1 - 1 531
          HQ122914.1 - 1 531
          Gene Symbol: POU5F1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001159542.1 NP_001153014.1 1 784
          GenBank mRNA AF268615.1 - 1 529
          AF268618.1 - 1 529
          DQ486513.1 - 1 784
          GU480835.1 - 1 530
          GU480840.1 - 1 530
          GU480842.1 - 1 530
          GU480845.1 - 1 530
          GU480849.1 - 1 532
          GU480850.1 - 1 531
          GU480864.1 - 1 530
          GU480868.1 - 1 530
          GU480878.1 - 1 530
          GU480880.1 - 1 530
          GU480883.1 - 1 530
          GU480884.1 - 1 529
          GU480885.1 - 1 531
          GU480886.1 - 1 530
          GU480889.1 - 1 530
          GU480899.1 - 1 531
          GU480902.1 - 1 530
          GU480905.1 - 1 530
          GU480906.1 - 1 530
          GU480908.1 - 1 530
          GU480911.1 - 1 530
          GU480913.1 - 1 530
          HQ122713.1 - 1 530
          HQ122714.1 - 1 530
          HQ122715.1 - 1 529
          HQ122716.1 - 1 530
          HQ122717.1 - 1 530
          HQ122718.1 - 1 530
          HQ122719.1 - 1 530
          HQ122720.1 - 1 530
          HQ122721.1 - 1 530
          HQ122750.1 - 1 530
          HQ122751.1 - 1 530
          HQ122752.1 - 1 530
          HQ122781.1 - 1 530
          HQ122782.1 - 1 530
          HQ122784.1 - 1 530
          HQ122785.1 - 1 530
          HQ122786.1 - 1 530
          HQ122787.1 - 1 530
          HQ122788.1 - 1 530
          HQ122789.1 - 1 530
          HQ122790.1 - 1 530
          HQ122816.1 - 1 530
          HQ122817.1 - 1 530
          HQ122824.1 - 1 530
          HQ122827.1 - 1 530
          HQ122841.1 - 1 530
          HQ122844.1 - 1 530
          HQ122852.1 - 1 530
          HQ122853.1 - 1 530
          HQ122856.1 - 1 530
          HQ122863.1 - 1 530
          HQ122864.1 - 1 530
          HQ122865.1 - 1 530
          HQ122866.1 - 1 530
          HQ122867.1 - 1 530
          HQ122868.1 - 1 530
          HQ122869.1 - 1 530
          HQ122870.1 - 1 530
          HQ122871.1 - 1 530
          HQ122872.1 - 1 529
          HQ122873.1 - 1 529
          HQ122874.1 - 1 530
          HQ122875.1 - 1 530
          HQ122876.1 - 1 530
          HQ122877.1 - 1 530
          HQ122878.1 - 1 530
          HQ122879.1 - 1 530
          HQ122880.1 - 1 530
          HQ122881.1 - 1 530
          HQ122882.1 - 1 530
          HQ122883.1 - 1 530
          HQ122884.1 - 1 530
          HQ122885.1 - 1 530
          HQ122886.1 - 1 530
          HQ122887.1 - 1 530
          HQ122891.1 - 1 530
          HQ122893.1 - 1 530
          HQ122894.1 - 1 530
          HQ122895.1 - 1 530
          HQ122896.1 - 1 530
          HQ122897.1 - 1 530
          HQ122898.1 - 1 530
          HQ122899.1 - 1 530
          HQ122900.1 - 1 530
          HQ122901.1 - 1 531
          HQ122903.1 - 1 530
          Gene Symbol: POU5F1P3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036440.1 1 528
          GenBank mRNA AF268617.1 - 1 527
          GU480832.1 - 1 528
          GU480837.1 - 1 528
          GU480838.1 - 1 528
          GU480843.1 - 1 528
          GU480846.1 - 1 528
          GU480847.1 - 1 528
          GU480876.1 - 1 528
          GU480877.1 - 1 528
          GU480887.1 - 1 528
          GU480888.1 - 1 529
          GU480891.1 - 1 529
          GU480892.1 - 1 528
          GU480893.1 - 1 528
          GU480897.1 - 1 528
          GU480901.1 - 1 527
          GU480903.1 - 1 528
          GU480907.1 - 1 528
          GU480909.1 - 1 528
          GU480915.1 - 1 528
          HQ122692.1 - 1 528
          HQ122693.1 - 1 528
          HQ122694.1 - 1 528
          HQ122722.1 - 1 528
          HQ122723.1 - 1 528
          HQ122724.1 - 1 528
          HQ122725.1 - 1 528
          HQ122726.1 - 1 528
          HQ122727.1 - 1 528
          HQ122728.1 - 1 528
          HQ122729.1 - 1 528
          HQ122730.1 - 1 528
          HQ122753.1 - 1 528
          HQ122754.1 - 1 528
          HQ122755.1 - 1 528
          HQ122791.1 - 1 528
          HQ122792.1 - 1 528
          HQ122793.1 - 1 528
          HQ122794.1 - 1 528
          HQ122795.1 - 1 528
          HQ122796.1 - 1 528
          HQ122797.1 - 1 528
          HQ122828.1 - 1 528
          HQ122829.1 - 1 528
          HQ122830.1 - 1 528
          HQ122831.1 - 1 528

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc