Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006983.1 | NP_008914.1 | 8 | 1127 | ||
XM_017002615.1 | XP_016858104.1 | 7 | 2035 | |||
XM_017002616.1 | XP_016858105.1 | 7 | 2032 | |||
GenBank mRNA | AB010961.1 | - | 8 | 1116 | ||
AB010962.1 | - | 7 | 1208 | |||
AF056200.1 | - | 8 | 1104 | |||
AJ005256.1 | - | 8 | 1127 | |||
BC025719.1 | - | 8 | 1118 | 5228894 |
Gene Symbol: MMP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002946.1 | 6 | 796 | |||
GenBank mRNA | AF057061.1 | - | 5 | 770 | ||
AF057062.1 | - | 4 | 722 | |||
AF086353.1 | - | 5 | 542 |