Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006983.1 | NP_008914.1 | 7 | 1010 | ||
XM_017002615.1 | XP_016858104.1 | 6 | 1918 | |||
XM_017002616.1 | XP_016858105.1 | 6 | 1918 | |||
XM_017002617.1 | XP_016858106.1 | 6 | 1918 | |||
GenBank mRNA | AB010961.1 | - | 7 | 999 | ||
AB010962.1 | - | 6 | 1091 | |||
AF056200.1 | - | 7 | 987 | |||
AJ005256.1 | - | 7 | 1010 | |||
AK297700.1 | - | 5 | 1482 | |||
BC025719.1 | - | 7 | 1001 | 5228894 |
Gene Symbol: MMP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002946.1 | 5 | 679 | |||
GenBank mRNA | AF057061.1 | - | 4 | 653 | ||
AF057062.1 | - | 3 | 605 | |||
AF086353.1 | - | 4 | 425 |