Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF518A SIRNA Assays ›
Gene Symbol:
ZNF518A
Gene Name:
zinc finger protein 518A
Gene Aliases:
ZNF518
Chromosome Location:
Chr.10: 96129220 - 96165512 on Build GRCh38
UniGene ID:
Hs.600823
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS145330
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF518A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001278524.1
NP_001265453.1
7
2989
NM_001278525.1
NP_001265454.1
6
2650
NM_001278526.1
NP_001265455.1
7
1274
NM_014803.3
NP_055618.2
6
2943
XM_011540406.2
XP_011538708.1
7
2802
XM_011540408.2
XP_011538710.1
6
3376
XM_011540410.2
XP_011538712.1
7
2824
XM_011540412.2
XP_011538714.1
6
3337
XM_011540413.2
XP_011538715.1
7
3462
XM_011540415.2
XP_011538717.1
6
2655
XM_011540418.2
XP_011538720.1
7
2771
XM_011540419.2
XP_011538721.1
5
2600
XM_011540420.2
XP_011538722.1
5
2627
XM_017016986.1
XP_016872475.1
7
3313
XM_017016987.1
XP_016872476.1
7
3340
XM_017016988.1
XP_016872477.1
7
3383
XM_017016989.1
XP_016872478.1
6
2678
XM_017016990.1
XP_016872479.1
6
3189
XM_017016991.1
XP_016872480.1
6
3162
XM_017016992.1
XP_016872481.1
7
3192
XM_017016993.1
XP_016872482.1
6
2673
XM_017016994.1
XP_016872483.1
6
3311
XM_017016995.1
XP_016872484.1
7
3216
XM_017016996.1
XP_016872485.1
7
3309
XM_017016997.1
XP_016872486.1
3
3522
XM_017016998.1
XP_016872487.1
2
3851
XM_017016999.1
XP_016872488.1
3
3304
GenBank mRNA
AB002333.2
-
2
2917
BC109045.1
-
7
1090
40028941
BC109046.1
-
7
1085
40028950
CR627437.1
-
6
2944
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0