Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320412.1 | NP_001307341.1 | 10, 11 | 1298 | ||
NM_006657.2 | NP_006648.1 | 10, 11 | 1298 | |||
NM_206965.1 | NP_996848.1 | 10, 11 | 1298 | |||
XM_011529435.2 | XP_011527737.1 | 10, 11 | 1596 | |||
GenBank mRNA | AF169017.1 | - | 10, 11 | 1298 | ||
AF289021.1 | - | 10, 11 | 1298 | |||
AF289022.1 | - | 10, 11 | 1298 | |||
AF289024.1 | - | 10, 11 | 1323 | |||
BC052248.1 | - | 10, 11 | 1294 | 6270063 | ||
BC136383.1 | - | 10, 11 | 1290 | 9020370 | ||
BC136395.1 | - | 10, 11 | 1290 | 9020382 | ||
BI966164.1 | - | 1, 2 | 136 | |||
BM021970.1 | - | 1, 2 | 133 | |||
U91541.1 | - | 1, 2 | 69 |