Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKFZP434L187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026771.1 | 3 | 1174 | |||
GenBank mRNA | AL117445.1 | - | 2 | 1064 | ||
BC130408.1 | - | 3 | 1174 | |||
BC130410.1 | - | 3 | 1174 |
Gene Symbol: ALDH16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK074136.1 | - | 15 | 3239 |
Gene Symbol: KLHL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250901.4 | XP_005250958.1 | 4 | 2545 |
Gene Symbol: LOC105375980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127963.1 | - | 10 | 1631 |
Gene Symbol: TMEM119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097290.1 | - | 4 | 843 | ||
BC096825.1 | - | 2 | 1259 | 30558500 |