Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IP6K2 SIRNA Assays ›
Gene Symbol:
IP6K2
Gene Name:
inositol hexakisphosphate kinase 2
Gene Aliases:
IHPK2, PIUS
Chromosome Location:
Chr.3: 48688003 - 48717278 on Build GRCh38
UniGene ID:
Hs.595983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS147459
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IP6K2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005909.2
NP_001005909.1
2
420
NM_001005910.2
NP_001005910.1
2
423
NM_001005911.2
NP_001005911.1
2
420
NM_001146178.2
NP_001139650.1
2
423
NM_001146179.2
NP_001139651.1
2
420
NM_001190316.1
NP_001177245.1
3
636
NM_001190317.1
NP_001177246.1
3
559
NM_016291.3
NP_057375.2
2
423
NR_027437.2
2
423
NR_027438.2
2
420
XM_006713199.3
XP_006713262.1
3
527
XM_006713200.1
XP_006713263.1
3
497
XM_006713201.1
XP_006713264.1
3
574
XM_006713202.1
XP_006713265.1
3
576
XM_011533816.1
XP_011532118.1
3
557
XM_011533817.1
XP_011532119.1
2
360
XM_011533818.1
XP_011532120.1
2
363
XM_011533822.1
XP_011532124.1
2
360
XM_011533823.1
XP_011532125.1
2
363
XM_017006583.1
XP_016862072.1
2
419
XM_017006584.1
XP_016862073.1
2
394
XM_017006585.1
XP_016862074.1
3
509
XM_017006586.1
XP_016862075.1
3
550
XM_017006587.1
XP_016862076.1
3
550
XM_017006588.1
XP_016862077.1
3
576
XM_017006589.1
XP_016862078.1
3
525
XM_017006590.1
XP_016862079.1
3
1131
XM_017006591.1
XP_016862080.1
3
496
XM_017006592.1
XP_016862081.1
3
525
XM_017006593.1
XP_016862082.1
3
525
GenBank mRNA
AF177145.1
-
1
317
AK024045.1
-
2
360
AK097016.1
-
8
1278
AK290526.1
-
3
574
AK291501.1
-
3
551
AK304714.1
-
3
497
AK315204.1
-
2
361
AL137514.1
-
2
361
AY007091.1
-
2
346
BC001864.1
-
2
345
BC004469.2
-
2
350
BC019694.1
-
2
332
4941646
BC065533.1
-
2
349
6162048
BP262706.1
-
2
360
BX537544.1
-
3
575
CA488567.1
-
2
357
CD248129.1
-
2
386
CF139656.1
-
2
354
CN362780.1
-
2
335
DA083858.1
-
2
356
DB039955.1
-
2
420
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
phosphate ion transport
negative regulation of cell growth
positive regulation of apoptotic process
inositol phosphate metabolic process
phosphatidylinositol phosphorylation
type I interferon signaling pathway
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
protein binding
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0