Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172105.1 | NP_001165576.1 | 12 | 1639 | ||
NM_017577.4 | NP_060047.3 | 17 | 1912 | |||
XM_005247546.2 | XP_005247603.1 | 16 | 1828 | |||
XM_005247547.1 | XP_005247604.1 | 16 | 1697 | |||
XM_011512930.1 | XP_011511232.1 | 17 | 1907 | |||
XM_011512931.1 | XP_011511233.1 | 12 | 1696 | |||
XM_017006646.1 | XP_016862135.1 | 15 | 1649 | |||
XM_017006647.1 | XP_016862136.1 | 18 | 1990 | |||
XM_017006648.1 | XP_016862137.1 | 13 | 1719 | |||
XM_017006649.1 | XP_016862138.1 | 11 | 1470 | |||
GenBank mRNA | AK093181.1 | - | 12 | 1431 | ||
AK097783.1 | - | 18 | 1996 | |||
AK292846.1 | - | 17 | 1848 | |||
AK292964.1 | - | 12 | 1554 | |||
AK304142.1 | - | 12 | 1639 | |||
AL133661.1 | - | 8 | 2128 | |||
AY358725.1 | - | 12 | 1592 | |||
BC028972.2 | - | 9 | 916 | |||
BC035040.1 | - | 17 | 1918 | 4829130 | ||
BX648633.1 | - | 1 | 1949 |