Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMBRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267782.1 | NP_001254711.1 | 20 | 4181 | ||
NM_001267783.1 | NP_001254712.1 | 18 | 3815 | |||
NM_001300731.1 | NP_001287660.1 | 17 | 3992 | |||
NM_017749.3 | NP_060219.2 | 19 | 3900 | |||
XM_005253009.3 | XP_005253066.1 | 18 | 4168 | |||
XM_005253011.3 | XP_005253068.1 | 17 | 4081 | |||
XM_005253014.3 | XP_005253071.1 | 19 | 3898 | |||
XM_006718259.2 | XP_006718322.1 | 18 | 4049 | |||
XM_006718260.2 | XP_006718323.1 | 17 | 3631 | |||
XM_017018007.1 | XP_016873496.1 | 16 | 3901 | |||
GenBank mRNA | AB051523.1 | - | 17 | 3944 | ||
AK000301.1 | - | 20 | 4131 | |||
AK023197.1 | - | 12 | 2423 | |||
AK057896.1 | - | 3 | 621 | |||
AK302808.1 | - | 18 | 4230 | |||
AL834190.1 | - | 18 | 3695 | |||
DQ870924.1 | - | 16 | 3723 |