Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN7L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098833.1 | NP_001092303.1 | 6, 7 | 625 | ||
NM_020218.1 | NP_064603.1 | 6, 7 | 646 | |||
XM_011525038.2 | XP_011523340.1 | 7 | 1297 | |||
XM_011525044.2 | XP_011523346.1 | 6, 7 | 1240 | |||
XM_011525046.2 | XP_011523348.1 | 6, 7 | 1219 | |||
XM_017024880.1 | XP_016880369.1 | 8 | 875 | |||
XM_017024881.1 | XP_016880370.1 | 8 | 1174 | |||
XM_017024882.1 | XP_016880371.1 | 7 | 1276 | |||
XM_017024883.1 | XP_016880372.1 | 8 | 878 | |||
XM_017024884.1 | XP_016880373.1 | 7 | 900 | |||
XM_017024885.1 | XP_016880374.1 | 7, 8 | 821 | |||
XM_017024886.1 | XP_016880375.1 | 7, 8 | 800 | |||
GenBank mRNA | AK056002.1 | - | 3, 4 | 169 | ||
AK126117.1 | - | 6, 7 | 843 | |||
AL390158.1 | - | 1, 2 | 55 | |||
BC037418.1 | - | 6, 7 | 553 | |||
BC113595.1 | - | 6, 7 | 625 | 8322647 | ||
BC126113.1 | - | 6, 7 | 625 | 8991829 |