Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIGYF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022574.4 | NP_072096.2 | 20 | 3458 | ||
XM_005250532.1 | XP_005250589.1 | 22 | 3239 | |||
XM_011516472.1 | XP_011514774.1 | 22 | 3239 | |||
XM_011516477.1 | XP_011514779.1 | 22 | 3236 | |||
XM_017012526.1 | XP_016868015.1 | 22 | 3239 | |||
XM_017012527.1 | XP_016868016.1 | 24 | 3137 | |||
XM_017012528.1 | XP_016868017.1 | 23 | 2749 | |||
XM_017012529.1 | XP_016868018.1 | 23 | 2756 | |||
XM_017012530.1 | XP_016868019.1 | 23 | 3010 | |||
XM_017012531.1 | XP_016868020.1 | 18 | 2262 | |||
GenBank mRNA | AF289554.1 | - | 2 | 265 | ||
AY176044.1 | - | 20 | 3659 | |||
BC018563.1 | - | 4 | 432 | |||
BC129991.1 | - | 14 | 1606 |