Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP2-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ406929.1 | - | 286 |
Gene Symbol: KRTAP2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033032.2 | NP_149021.2 | 1 | 465 | ||
GenBank mRNA | AJ302536.1 | - | 352 |