Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301834.1 | NP_001288763.1 | 2 | 212 | ||
NM_001301837.1 | NP_001288766.1 | 1 | 341 | |||
NM_001301838.1 | NP_001288767.1 | 1 | 341 | |||
NM_138425.3 | NP_612434.1 | 1 | 341 | |||
NR_126035.1 | 1 | 341 | ||||
GenBank mRNA | AK311912.1 | - | 1 | 139 | ||
AV751928.1 | - | 83 | ||||
BC009925.2 | - | 1 | 118 | 3537206 | ||
BE888740.1 | - | 1 | 32 | |||
BM687523.1 | - | 1 | 328 | |||
BM781587.1 | - | 1 | 341 | |||
CB161655.1 | - | 2 | 212 |