Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other XRRA1 SIRNA Assays ›
Gene Symbol:
XRRA1
Gene Name:
X-ray radiation resistance associated 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 74840910 - 74949187 on Build GRCh38
UniGene ID:
Hs.370145
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS152055
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: XRRA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270380.1
NP_001257309.1
12
1548
NM_001270381.1
NP_001257310.1
12
1541
NM_182969.2
NP_892014.1
15
1809
XM_005273765.2
XP_005273822.1
15
1833
XM_005273770.2
XP_005273827.1
13
1613
XM_011544755.1
XP_011543057.1
15
1788
XM_011544756.1
XP_011543058.1
14
1707
XM_011544757.1
XP_011543059.1
14
1662
XM_011544758.1
XP_011543060.1
13
1662
XM_011544759.1
XP_011543061.1
13
1638
XM_011544760.2
XP_011543062.1
15
1833
XM_011544761.1
XP_011543063.1
12
1590
XM_011544762.1
XP_011543064.1
12
1572
XM_011544763.1
XP_011543065.1
15
1833
XM_011544764.1
XP_011543066.1
12
1497
XM_011544765.1
XP_011543067.1
13
1547
XM_011544766.2
XP_011543068.1
15
1852
XM_011544767.1
XP_011543069.1
14
1754
XM_011544768.1
XP_011543070.1
13
1601
XM_017017221.1
XP_016872710.1
13
1623
XM_017017222.1
XP_016872711.1
14
1707
XM_017017223.1
XP_016872712.1
13
1662
XM_017017224.1
XP_016872713.1
12
1572
XM_017017225.1
XP_016872714.1
12
1497
XM_017017226.1
XP_016872715.1
14
1728
XM_017017227.1
XP_016872716.1
14
1704
XM_017017228.1
XP_016872717.1
13
1613
XM_017017229.1
XP_016872718.1
13
1557
XM_017017230.1
XP_016872719.1
12
1381
XM_017017231.1
XP_016872720.1
11
1467
XM_017017232.1
XP_016872721.1
10
1277
XM_017017233.1
XP_016872722.1
12
1561
XM_017017234.1
XP_016872723.1
11
1446
GenBank mRNA
AF439934.1
-
4
434
AK056364.1
-
10
1132
AK074152.1
-
10
1139
AK091449.1
-
2
1255
BC037294.2
-
12
1474
BC040541.1
-
12
1474
BC064584.1
-
12
1477
5742492
CR749605.1
-
11
1513
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to X-ray
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline