Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM161B SIRNA Assays ›
Gene Symbol:
TMEM161B
Gene Name:
transmembrane protein 161B
Gene Aliases:
FLB3342, PRO1313
Chromosome Location:
Chr.5: 88189419 - 88268917 on Build GRCh38
UniGene ID:
Hs.379972
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS152736
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM161B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289007.1
NP_001275936.1
10
1156
NM_001289008.1
NP_001275937.1
11
1288
NM_153354.4
NP_699185.1
10
1156
NR_110266.1
11
1256
XM_006714553.1
XP_006714616.1
10
1106
XM_006714554.3
XP_006714617.1
9
1043
XM_006714555.3
XP_006714618.1
8
925
XM_006714556.3
XP_006714619.2
8
886
XM_011543201.2
XP_011541503.1
9
1023
XM_011543202.2
XP_011541504.2
8
1568
XM_011543203.2
XP_011541505.2
6
1373
XM_011543204.2
XP_011541506.2
8
841
XM_017009093.1
XP_016864582.1
10
1106
XM_017009094.1
XP_016864583.1
9
997
XM_017009095.1
XP_016864584.1
10
1106
XM_017009096.1
XP_016864585.1
9
1027
XM_017009097.1
XP_016864586.1
9
1030
XM_017009098.1
XP_016864587.1
9
975
XM_017009099.1
XP_016864588.1
8
904
XM_017009100.1
XP_016864589.1
9
997
XM_017009101.1
XP_016864590.1
9
959
XM_017009102.1
XP_016864591.1
9
960
XM_017009103.1
XP_016864592.1
9
1027
XM_017009104.1
XP_016864593.1
9
1090
XM_017009105.1
XP_016864594.1
10
1068
XM_017009106.1
XP_016864595.1
9
960
XM_017009107.1
XP_016864596.1
9
990
XM_017009108.1
XP_016864597.1
8
875
XM_017009109.1
XP_016864598.1
9
1040
XM_017009110.1
XP_016864599.1
9
1026
GenBank mRNA
AK300893.1
-
11
1238
AK301091.1
-
10
1107
BC037287.2
-
10
1125
4837154
BM479365.1
-
7
765
BX647345.1
-
11
1176
BX648757.1
-
8
4929
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix