Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271049.1 | NP_001257978.1 | 13 | 1380 | ||
NR_073132.1 | 14 | 1525 | ||||
NR_073133.1 | 12 | 1279 | ||||
XM_006712353.3 | XP_006712416.1 | 13 | 1420 | |||
XM_017003550.1 | XP_016859039.1 | 13 | 1420 | |||
XM_017003551.1 | XP_016859040.1 | 13 | 1547 | |||
XM_017003552.1 | XP_016859041.1 | 13 | 1433 | |||
XM_017003553.1 | XP_016859042.1 | 13 | 1766 | |||
XM_017003554.1 | XP_016859043.1 | 12 | 1262 | |||
XM_017003555.1 | XP_016859044.1 | 12 | 1262 | |||
XM_017003556.1 | XP_016859045.1 | 12 | 1265 | |||
XM_017003557.1 | XP_016859046.1 | 11 | 1195 | |||
XM_017003558.1 | XP_016859047.1 | 13 | 1420 | |||
XM_017003559.1 | XP_016859048.1 | 10 | 2391 | |||
GenBank mRNA | AK293665.1 | - | 12 | 1262 | ||
BC036530.1 | - | 14 | 1491 | 4828213 |