Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM133A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171109.1 | NP_001164580.1 | 4 | 615 | ||
NM_001171110.1 | NP_001164581.1 | 5 | 650 | |||
NM_001171111.1 | NP_001164582.1 | 3 | 526 | |||
NM_173698.2 | NP_775969.1 | 4 | 595 | |||
XM_006724641.3 | XP_006724704.1 | 3 | 627 | |||
XM_011530925.2 | XP_011529227.1 | 4 | 568 | |||
XM_011530926.2 | XP_011529228.1 | 4 | 406 | |||
XM_011530927.2 | XP_011529229.1 | 4 | 445 | |||
XM_011530928.1 | XP_011529230.1 | 4 | 379 | |||
XM_011530929.1 | XP_011529231.1 | 5 | 414 | |||
XM_011530931.2 | XP_011529233.1 | 3 | 581 | |||
XM_017029454.1 | XP_016884943.1 | 5 | 3254 | |||
XM_017029455.1 | XP_016884944.1 | 6 | 587 | |||
XM_017029456.1 | XP_016884945.1 | 4 | 522 | |||
GenBank mRNA | AK094978.1 | - | 4 | 402 | ||
BC113385.1 | - | 1 | 112 | 8322437 | ||
BC113387.1 | - | 1 | 112 | 8322439 | ||
DA188870.1 | - | 5 | 460 | |||
DA297338.1 | - | 4 | 402 | |||
DA414074.1 | - | 4 | 425 | |||
DB084469.1 | - | 3 | 337 |