Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103908605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126050.1 | 1 | 118 | |||
GenBank mRNA | DA585556.1 | - | 1 | 118 |
Gene Symbol: GLIDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015363.2 | 1 | 226 | |||
NR_126044.1 | 1 | 226 | ||||
NR_126045.1 | 1 | 226 | ||||
NR_126046.1 | 1 | 226 | ||||
GenBank mRNA | AK092305.1 | - | 1 | 125 | ||
AK095690.1 | - | 1 | 127 | |||
BC027338.1 | - | 1 | 130 |
Gene Symbol: KGFLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003674.2 | 1 | 442 | |||
NR_047527.1 | 1 | 442 | ||||
GenBank mRNA | AF523265.1 | - | 1 | 537 | ||
AK054997.1 | - | 1 | 181 | |||
AK307215.1 | - | 1 | 173 | |||
AY098593.1 | - | 1 | 433 | |||
BC101842.1 | - | 1 | 442 | |||
BC112977.1 | - | 1 | 440 | |||
BC112978.1 | - | 1 | 440 |
Gene Symbol: KGFLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003670.1 | 1 | 541 | |||
GenBank mRNA | AY098594.1 | - | 1 | 433 | ||
BC143690.1 | - | 1 | 443 |
Gene Symbol: LOC100126582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296330.1 | - | 1 | 448 | ||
BC108270.1 | - | 1 | 537 | 4776745 |
Gene Symbol: LOC102724238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126048.1 | 1 | 170 | |||
GenBank mRNA | AK290223.1 | - | 1 | 449 | ||
AK292642.1 | - | 1 | 97 |
Gene Symbol: LOC103908604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA742734.1 | - | 1 | 145 |
Gene Symbol: LOC554249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126049.1 | 1 | 166 | |||
GenBank mRNA | BC084570.1 | - | 1 | 80 | 6378300 |