Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013643.1 | NP_001013665.1 | 2, 3 | 181 | ||
GenBank mRNA | AA922397.1 | - | 2, 3 | 201 |