Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101926934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134643.1 | 1 | 127 | |||
GenBank mRNA | AI698692.1 | - | 2 | 241 |
Gene Symbol: TDRD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010870.2 | NP_001010870.1 | 1 | 161 | ||
NM_001168359.1 | NP_001161831.1 | 1 | 161 | |||
GenBank mRNA | AK131455.1 | - | 1 | 161 |