Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN3 SIRNA Assays ›
Gene Symbol:
RTN3
Gene Name:
reticulon 3
Gene Aliases:
ASYIP, HAP, NSPL2, NSPLII, RTN3-A1
Chromosome Location:
Chr.11: 63681315 - 63759891 on Build GRCh38
UniGene ID:
Hs.743229
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS173552
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001265589.1
NP_001252518.1
4
2782
NM_001265590.1
NP_001252519.1
3
2446
NM_001265591.1
NP_001252520.1
2
394
NM_006054.3
NP_006045.1
2
394
NM_201428.2
NP_958831.1
3
2725
NM_201429.2
NP_958832.1
3
451
NM_201430.2
NP_958833.1
2
394
NR_049750.1
2
394
NR_049751.1
2
394
XM_011544730.2
XP_011543032.1
4
2872
XM_011544731.2
XP_011543033.1
3
586
GenBank mRNA
AB209771.1
-
2
331
AF059524.1
-
2
331
AF119297.1
-
2
343
AK075412.1
-
2
344
AK094965.1
-
2
1447
AK122727.1
-
2
847
AK127079.1
-
3
2421
AK222898.1
-
2
331
AK295361.1
-
3
356
AK295534.1
-
2
248
AK295758.1
-
3
2374
AK297529.1
-
2
271
AK297533.1
-
2
344
AK304100.1
-
4
2732
AK307811.1
-
3
2671
AK308083.1
-
2
339
AK309766.1
-
3
2400
AY427821.2
-
3
2692
AY750848.1
-
4
2749
BC000634.1
-
2
302
BC010556.1
-
2
307
BC011394.1
-
2
343
BC022993.1
-
2
333
BC100822.2
-
2
324
BC100823.2
-
2
324
BC105981.1
-
2
275
BC105982.1
-
2
306
BC118550.1
-
2
324
BC118628.1
-
2
324
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
chemical synaptic transmission
viral process
vesicle-mediated transport
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0