Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081442.2 | NP_001074911.1 | 3 | 380 | ||
NM_001081443.2 | NP_001074912.1 | 3 | 380 | |||
NM_001304457.1 | NP_001291386.1 | 3 | 380 | |||
NM_006840.4 | NP_006831.1 | 3 | 380 | |||
XM_011526359.2 | XP_011524661.1 | 3 | 364 | |||
XM_011526360.2 | XP_011524662.1 | 3 | 364 | |||
XM_011526361.2 | XP_011524663.1 | 3 | 364 | |||
XM_011526362.2 | XP_011524664.1 | 3 | 345 | |||
GenBank mRNA | AF025534.1 | - | 3 | 331 | ||
AK057072.1 | - | 3 | 334 | |||
AK090399.1 | - | 1 | 667 | |||
AK092867.1 | - | 3 | 248 | |||
AK223296.1 | - | 3 | 336 | |||
AK290845.1 | - | 3 | 364 | |||
AK301112.1 | - | 3 | 343 | |||
BC025704.1 | - | 3 | 334 | 5223581 | ||
DC320482.1 | - | 3 | 380 |