Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195639.1 | NP_001182568.1 | 6 | 1103 | ||
NM_052958.2 | NP_443190.2 | 6 | 1103 | |||
XM_011517445.2 | XP_011515747.1 | 6 | 1075 | |||
XM_011517446.2 | XP_011515748.1 | 6 | 1075 | |||
XM_011517448.2 | XP_011515750.1 | 5 | 946 | |||
XM_011517449.2 | XP_011515751.1 | 6 | 1075 | |||
XM_011517450.2 | XP_011515752.1 | 6 | 1075 | |||
XM_017013019.1 | XP_016868508.1 | 5 | 946 | |||
GenBank mRNA | AB056652.1 | - | 5 | 2071 | ||
AK291436.1 | - | 6 | 575 | |||
AL834454.1 | - | 6 | 600 | |||
BC041961.1 | - | 6 | 1103 | 5302156 |