Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM150A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031738.2 | NP_001026908.1 | 3, 4 | 316 | ||
NR_033179.1 | 3, 4 | 316 | ||||
XM_006711930.3 | XP_006711993.1 | 3, 4 | 516 | |||
XM_006711931.3 | XP_006711994.1 | 3, 4 | 355 | |||
XM_006711932.2 | XP_006711995.1 | 2, 3 | 553 | |||
XM_011532505.2 | XP_011530807.1 | 2, 3 | 351 | |||
XM_011532508.2 | XP_011530810.1 | 2, 3 | 351 | |||
GenBank mRNA | AK074505.1 | - | 3, 4 | 317 | ||
AK291879.1 | - | 3, 4 | 282 | |||
BC050466.1 | - | 3, 4 | 276 | 6197700 | ||
BX647409.1 | - | 3, 4 | 252 | |||
DA434376.1 | - | 3, 4 | 317 |